N-Ethyl-N-(4-propoxyphenyl)amine

Supplier: Matrix Scientific

031899-500MG
101832-622EA 338.46 USD
101832-622
N-Ethyl-N-(4-propoxyphenyl)amine
N-Ethyl-N-(4-propoxyphenyl)amine

Hazard: Irritant

Formula: C₁₁H₁₇NO
Boiling Pt: 145…147 °C (13 Torr)
MDL Number: MFCD10687915
CAS Number: 59731-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR