N-[2-(4-Chloro-2-methylphenoxy)ethyl]-4-propoxyaniline

Supplier: Matrix Scientific

031896-500MG
101832-616EA 306.15 USD
101832-616
N-[2-(4-Chloro-2-methylphenoxy)ethyl]-4-propoxyaniline
N-[2-(4-Chloro-2-methylphenoxy)ethyl]-4-propoxyaniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO₂
MDL Number: MFCD10687912
CAS Number: 1040688-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR