N-[2-(4-Chlorophenoxy)ethyl]-4-propoxyaniline

Supplier: Matrix Scientific

031895-500MG
101832-614EA 306.15 USD
101832-614
N-[2-(4-Chlorophenoxy)ethyl]-4-propoxyaniline
N-[2-(4-Chlorophenoxy)ethyl]-4-propoxyaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO₂
MDL Number: MFCD10687911
CAS Number: 1040688-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR