4-Propoxy-N-(2-thienylmethyl)aniline

Supplier: Matrix Scientific

031894-500MG
101832-612EA 306.15 USD
101832-612
4-Propoxy-N-(2-thienylmethyl)aniline
4-Propoxy-N-(2-thienylmethyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₇NOS
MDL Number: MFCD10687910
CAS Number: 1040688-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR