2-Chloro-N-(cyclohexylmethyl)aniline

Supplier: Matrix Scientific

031891-500MG
101832-606EA 306.15 USD
101832-606
2-Chloro-N-(cyclohexylmethyl)aniline
2-Chloro-N-(cyclohexylmethyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₈ClN
MDL Number: MFCD03211352
CAS Number: 130708-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR