4-Isobutoxy-N-[2-(4-isopropylphenoxy)propyl]aniline

Supplier: Matrix Scientific

031888-500MG
101832-600EA 306.15 USD
101832-600
4-Isobutoxy-N-[2-(4-isopropylphenoxy)propyl]aniline
4-Isobutoxy-N-[2-(4-isopropylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₂₂H₃₁NO₂
MW: 341.5 g/mol
MDL Number: MFCD10687905
CAS Number: 1040688-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR