N-[2-(Hexyloxy)benzyl]-4-isobutoxyaniline

Supplier: Matrix Scientific

031886-500MG
101832-596EA 306.15 USD
101832-596
N-[2-(Hexyloxy)benzyl]-4-isobutoxyaniline
N-[2-(Hexyloxy)benzyl]-4-isobutoxyaniline

Hazard: Irritant

Formula: C₂₃H₃₃NO₂
MDL Number: MFCD10687903
CAS Number: 1040688-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR