4-Isobutoxy-N-(4-methoxyphenethyl)aniline

Supplier: Matrix Scientific

031883-500MG
101832-590EA 306.15 USD
101832-590
4-Isobutoxy-N-(4-methoxyphenethyl)aniline
4-Isobutoxy-N-(4-methoxyphenethyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687900
CAS Number: 1040687-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR