4-Isobutoxy-N-(3-phenylpropyl)aniline

Supplier: Matrix Scientific

031881-500MG
101832-586EA 306.15 USD
101832-586
4-Isobutoxy-N-(3-phenylpropyl)aniline
4-Isobutoxy-N-(3-phenylpropyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO
MDL Number: MFCD10687898
CAS Number: 1040687-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR