N-(2-Chlorobenzyl)-4-isobutoxyaniline

Supplier: Matrix Scientific

031880-500MG
101832-584EA 306.15 USD
101832-584
N-(2-Chlorobenzyl)-4-isobutoxyaniline
N-(2-Chlorobenzyl)-4-isobutoxyaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10687897
CAS Number: 1040687-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR