N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-isobutoxyaniline

Supplier: Matrix Scientific

031879-500MG
101832-582EA 306.15 USD
101832-582
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-isobutoxyaniline
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-isobutoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₆ClNO₂
MDL Number: MFCD10687896
CAS Number: 1040687-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR