4-Butoxy-N-(3,5-dichlorobenzyl)aniline
Supplier: Matrix Scientific
031877-500MG
101832-578EA
306.15
USD
101832-578
4-Butoxy-N-(3,5-dichlorobenzyl)aniline
4-Butoxy-N-(3,5-dichlorobenzyl)aniline
Hazard: Irritant
Formula:
C₁₇H₁₉Cl₂NO |
MDL Number:
MFCD10687894 CAS Number: 1040687-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...