4-Butoxy-N-[4-(isopentyloxy)benzyl]aniline

Supplier: Matrix Scientific

031875-500MG
101832-574EA 306.15 USD
101832-574
4-Butoxy-N-[4-(isopentyloxy)benzyl]aniline
4-Butoxy-N-[4-(isopentyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₂H₃₁NO₂
MDL Number: MFCD10687892
CAS Number: 1040687-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR