4-(Hexyloxy)-N-(3-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

031869-500MG
101832-562EA 306.15 USD
101832-562
4-(Hexyloxy)-N-(3-isopropoxybenzyl)aniline
4-(Hexyloxy)-N-(3-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₂H₃₁NO₂
MDL Number: MFCD10687887
CAS Number: 1040687-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR