4-(Hexyloxy)-N-[4-(2-phenoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

031868-500MG
101832-560EA 306.15 USD
101832-560
4-(Hexyloxy)-N-[4-(2-phenoxyethoxy)benzyl]aniline
4-(Hexyloxy)-N-[4-(2-phenoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₇H₃₃NO₃
MDL Number: MFCD10687886
CAS Number: 1040687-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR