N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-(hexyloxy)aniline

Supplier: Matrix Scientific

031865-500MG
101832-554EA 306.15 USD
101832-554
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-(hexyloxy)aniline
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-4-(hexyloxy)aniline

Hazard: Irritant

Formula: C₂₂H₃₀ClNO₂
MDL Number: MFCD10687883
CAS Number: 1040687-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR