N-([1,1'-Biphenyl]-4-ylmethyl)-2-chloroaniline

Supplier: Matrix Scientific

031864-500MG
101832-552EA 306.15 USD
101832-552
N-([1,1'-Biphenyl]-4-ylmethyl)-2-chloroaniline
N-([1,1'-Biphenyl]-4-ylmethyl)-2-chloroaniline

Hazard: Irritant

Formula: C₁₉H₁₆ClN
MDL Number: MFCD03210581
CAS Number: 356530-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR