N-(2-Thienylmethyl)-1-octanamine

Supplier: Matrix Scientific

031863-500MG
101832-550EA 306.15 USD
101832-550
N-(2-Thienylmethyl)-1-octanamine
N-(2-Thienylmethyl)-1-octanamine

Hazard: Irritant

Formula: C₁₃H₂₃NS
MDL Number: MFCD07410553
CAS Number: 855379-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR