N-(3-Ethoxybenzyl)(2-thienyl)methanamine

Supplier: Matrix Scientific

031862-500MG
101832-548EA 306.15 USD
101832-548
N-(3-Ethoxybenzyl)(2-thienyl)methanamine
N-(3-Ethoxybenzyl)(2-thienyl)methanamine

Hazard: Irritant

Formula: C₁₄H₁₇NOS
MDL Number: MFCD07405776
CAS Number: 893579-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR