N-(4-Isopropylbenzyl)aniline

Supplier: Matrix Scientific

031860-500MG
101832-544EA 306.15 USD
101832-544
N-(4-Isopropylbenzyl)aniline
N-(4-Isopropylbenzyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₉N
MDL Number: MFCD04564006
CAS Number: 81308-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR