N-[4-(2-Ethoxyethoxy)benzyl]cyclopentanamine

Supplier: Matrix Scientific

031854-500MG
101832-532EA 306.15 USD
101832-532
N-[4-(2-Ethoxyethoxy)benzyl]cyclopentanamine
N-[4-(2-Ethoxyethoxy)benzyl]cyclopentanamine

Hazard: Irritant

Formula: C₁₆H₂₅NO₂
MDL Number: MFCD10687877
CAS Number: 1040686-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR