N-Cyclopentyl-N-hexylamine

Supplier: Matrix Scientific

031852-500MG
101832-528EA 306.15 USD
101832-528
N-Cyclopentyl-N-hexylamine
N-Cyclopentyl-N-hexylamine

Hazard: Irritant

Formula: C₁₁H₂₃N
MDL Number: MFCD10687875
CAS Number: 20007-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR