N-[2-(4-Chlorophenoxy)propyl]cyclopentanamine

Supplier: Matrix Scientific

031851-500MG
101832-526EA 306.15 USD
101832-526
N-[2-(4-Chlorophenoxy)propyl]cyclopentanamine
N-[2-(4-Chlorophenoxy)propyl]cyclopentanamine

Hazard: Irritant

Formula: C₁₄H₂₀ClNO
MDL Number: MFCD10687874
CAS Number: 1040686-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR