N-Methyl-2-(2-methylphenyl)-1-ethanamine hydrochloride

Supplier: Matrix Scientific

031848-500MG
101832-520EA 306.15 USD
101832-520
N-Methyl-2-(2-methylphenyl)-1-ethanamine hydrochloride
N-Methyl-2-(2-methylphenyl)-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClN
Melting Pt: 167 °C
MDL Number: MFCD18384826
CAS Number: 780791-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR