(3-Ethoxyphenyl)-N-methylmethanamine

Supplier: Matrix Scientific

031846-500MG
101832-516EA 461.54 USD
101832-516
(3-Ethoxyphenyl)-N-methylmethanamine
(3-Ethoxyphenyl)-N-methylmethanamine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD07405834
CAS Number: 893581-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR