(2-Isobutoxyphenyl)-N-methylmethanamine

Supplier: Matrix Scientific

031845-500MG
101832-514EA 306.15 USD
101832-514
(2-Isobutoxyphenyl)-N-methylmethanamine
(2-Isobutoxyphenyl)-N-methylmethanamine

Hazard: Irritant

Formula: C₁₂H₁₉NO
MDL Number: MFCD08691889
CAS Number: 915921-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR