N-[4-(sec-Butoxy)benzyl]-1-ethanamine

Supplier: Matrix Scientific

031839-500MG
101832-502EA 306.15 USD
101832-502
N-[4-(sec-Butoxy)benzyl]-1-ethanamine
N-[4-(sec-Butoxy)benzyl]-1-ethanamine

Hazard: Irritant

Formula: C₁₃H₂₁NO
MDL Number: MFCD10687867
CAS Number: 1040686-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR