N-Ethyl-2-(4-isopropylphenoxy)-1-propanamine

Supplier: Matrix Scientific

031835-500MG
101832-494EA 306.15 USD
101832-494
N-Ethyl-2-(4-isopropylphenoxy)-1-propanamine
N-Ethyl-2-(4-isopropylphenoxy)-1-propanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687863
CAS Number: 1040686-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR