N-[4-(Isopentyloxy)benzyl]-1-ethanamine

Supplier: Matrix Scientific

031834-500MG
101832-492EA 306.15 USD
101832-492
N-[4-(Isopentyloxy)benzyl]-1-ethanamine
N-[4-(Isopentyloxy)benzyl]-1-ethanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687862
CAS Number: 23014-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR