N-[2-(Pentyloxy)benzyl]-1-ethanamine

Supplier: Matrix Scientific

031832-500MG
101832-488EA 306.15 USD
101832-488
N-[2-(Pentyloxy)benzyl]-1-ethanamine
N-[2-(Pentyloxy)benzyl]-1-ethanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687860
CAS Number: 1040686-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR