2-(4-Chloro-3,5-dimethylphenoxy)-N-ethyl-1-ethanamine

Supplier: Matrix Scientific

031827-500MG
101832-478EA 306.15 USD
101832-478
2-(4-Chloro-3,5-dimethylphenoxy)-N-ethyl-1-ethanamine
2-(4-Chloro-3,5-dimethylphenoxy)-N-ethyl-1-ethanamine

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD10687856
CAS Number: 1040685-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR