3-Chloro-4-methoxy-N-[2-(phenethyloxy)benzyl]aniline

Supplier: Matrix Scientific

031826-500MG
101832-476EA 306.15 USD
101832-476
3-Chloro-4-methoxy-N-[2-(phenethyloxy)benzyl]aniline
3-Chloro-4-methoxy-N-[2-(phenethyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₂H₂₂ClNO₂
MW: 367.88 g/mol
MDL Number: MFCD10687855
CAS Number: 1040685-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR