3-Chloro-N-[2-(4-isopropylphenoxy)propyl]-4-methoxyaniline

Supplier: Matrix Scientific

031824-500MG
101832-472EA 306.15 USD
101832-472
3-Chloro-N-[2-(4-isopropylphenoxy)propyl]-4-methoxyaniline
3-Chloro-N-[2-(4-isopropylphenoxy)propyl]-4-methoxyaniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO₂
MDL Number: MFCD10687853
CAS Number: 1040685-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR