N-(2-Butoxybenzyl)-3-chloro-4-methoxyaniline

Supplier: Matrix Scientific

031823-500MG
101832-470EA 306.15 USD
101832-470
N-(2-Butoxybenzyl)-3-chloro-4-methoxyaniline
N-(2-Butoxybenzyl)-3-chloro-4-methoxyaniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO₂
MDL Number: MFCD10687852
CAS Number: 1040685-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR