3-Chloro-4-methoxy-N-[2-(3-methylphenoxy)propyl]aniline

Supplier: Matrix Scientific

031820-500MG
101832-464EA 306.15 USD
101832-464
3-Chloro-4-methoxy-N-[2-(3-methylphenoxy)propyl]aniline
3-Chloro-4-methoxy-N-[2-(3-methylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO₂
MW: 305.81 g/mol
MDL Number: MFCD10687849
CAS Number: 1040685-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR