3-Chloro-4-methoxy-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

031818-500MG
101832-460EA 306.15 USD
101832-460
3-Chloro-4-methoxy-N-(3-methylbenzyl)aniline
3-Chloro-4-methoxy-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD10687847
CAS Number: 1019526-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR