5-Chloro-N-(3-isopropoxybenzyl)-2-methylaniline

Supplier: Matrix Scientific

031816-500MG
101832-456EA 306.15 USD
101832-456
5-Chloro-N-(3-isopropoxybenzyl)-2-methylaniline
5-Chloro-N-(3-isopropoxybenzyl)-2-methylaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10687845
CAS Number: 1040685-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR