N-(3-Butoxybenzyl)-5-chloro-2-methylaniline

Supplier: Matrix Scientific

031813-500MG
101832-450EA 306.15 USD
101832-450
N-(3-Butoxybenzyl)-5-chloro-2-methylaniline
N-(3-Butoxybenzyl)-5-chloro-2-methylaniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD10687842
CAS Number: 1040685-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR