5-Chloro-N-[2-(4-chlorophenoxy)propyl]-2-methylaniline

Supplier: Matrix Scientific

031811-500MG
101832-446EA 306.15 USD
101832-446
5-Chloro-N-[2-(4-chlorophenoxy)propyl]-2-methylaniline
5-Chloro-N-[2-(4-chlorophenoxy)propyl]-2-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO
MDL Number: MFCD10687840
CAS Number: 1040685-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR