5-Chloro-N-[2-(4-methoxyphenoxy)propyl]-2-methylaniline

Supplier: Matrix Scientific

031809-500MG
101832-442EA 306.15 USD
101832-442
5-Chloro-N-[2-(4-methoxyphenoxy)propyl]-2-methylaniline
5-Chloro-N-[2-(4-methoxyphenoxy)propyl]-2-methylaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO₂
MDL Number: MFCD10687838
CAS Number: 1040684-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR