N-(5-Chloro-2-methylphenyl)-N-[2-(3-methylphenoxy)propyl]amine

Supplier: Matrix Scientific

031808-500MG
101832-440EA 306.15 USD
101832-440
N-(5-Chloro-2-methylphenyl)-N-[2-(3-methylphenoxy)propyl]amine
N-(5-Chloro-2-methylphenyl)-N-[2-(3-methylphenoxy)propyl]amine

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MW: 289.81 g/mol
MDL Number: MFCD10687837
CAS Number: 1040684-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR