5-Chloro-N-(cyclohexylmethyl)-2-methylaniline

Supplier: Matrix Scientific

031807-500MG
101832-438EA 306.15 USD
101832-438
5-Chloro-N-(cyclohexylmethyl)-2-methylaniline
5-Chloro-N-(cyclohexylmethyl)-2-methylaniline

Hazard: Irritant

Formula: C₁₄H₂₀ClN
MDL Number: MFCD10687836
CAS Number: 1036485-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR