5-Chloro-2-methyl-N-phenethylaniline

Supplier: Matrix Scientific

031806-500MG
101832-436EA 306.15 USD
101832-436
5-Chloro-2-methyl-N-phenethylaniline
5-Chloro-2-methyl-N-phenethylaniline

Hazard: Irritant

Formula: C₁₅H₁₆ClN
MDL Number: MFCD10687835
CAS Number: 1040684-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR