N-Benzyl-5-chloro-2-methylaniline

Supplier: Matrix Scientific

031804-500MG
101832-432EA 306.15 USD
101832-432
N-Benzyl-5-chloro-2-methylaniline
N-Benzyl-5-chloro-2-methylaniline

Hazard: Irritant

Formula: C₁₄H₁₄ClN
MDL Number: MFCD10687833
CAS Number: 946666-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR