2-Ethyl-N-(3-isopropoxybenzyl)-6-methylaniline
Supplier: Matrix Scientific
031803-500MG
101832-430EA
306.15
USD
101832-430
2-Ethyl-N-(3-isopropoxybenzyl)-6-methylaniline
2-Ethyl-N-(3-isopropoxybenzyl)-6-methylaniline
Hazard: Irritant
Formula:
C₁₉H₂₅NO |
MDL Number:
MFCD10687832 CAS Number: 1040684-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...