N-[2-(2,6-Dimethylphenoxy)propyl]-2-ethyl-6-methylaniline
Supplier: Matrix Scientific
031802-500MG
101832-428EA
306.15
USD
101832-428
N-[2-(2,6-Dimethylphenoxy)propyl]-2-ethyl-6-methylaniline
N-[2-(2,6-Dimethylphenoxy)propyl]-2-ethyl-6-methylaniline
Hazard: Irritant
Formula:
C₂₀H₂₇NO |
MDL Number:
MFCD10687831 CAS Number: 1040684-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...