2-Ethyl-6-methyl-N-[2-(3-methylphenoxy)propyl]aniline

Supplier: Matrix Scientific

031799-500MG
101832-422EA 306.15 USD
101832-422
2-Ethyl-6-methyl-N-[2-(3-methylphenoxy)propyl]aniline
2-Ethyl-6-methyl-N-[2-(3-methylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO
MW: 283.42 g/mol
MDL Number: MFCD10687828
CAS Number: 1040684-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR