2-Ethyl-6-methyl-N-[2-(4-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031797-500MG
101832-418EA 306.15 USD
101832-418
2-Ethyl-6-methyl-N-[2-(4-methylphenoxy)ethyl]aniline
2-Ethyl-6-methyl-N-[2-(4-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO
MW: 269.39 g/mol
MDL Number: MFCD10687826
CAS Number: 1040684-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR