N-(2,4-Dichlorobenzyl)-2-ethyl-6-methylaniline

Supplier: Matrix Scientific

031796-500MG
101832-416EA 306.15 USD
101832-416
N-(2,4-Dichlorobenzyl)-2-ethyl-6-methylaniline
N-(2,4-Dichlorobenzyl)-2-ethyl-6-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂N
MDL Number: MFCD10687825
CAS Number: 1040684-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR