2-Ethyl-N-[2-(4-isopropylphenoxy)ethyl]-6-methylaniline

Supplier: Matrix Scientific

031795-500MG
101832-414EA 306.15 USD
101832-414
2-Ethyl-N-[2-(4-isopropylphenoxy)ethyl]-6-methylaniline
2-Ethyl-N-[2-(4-isopropylphenoxy)ethyl]-6-methylaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO
MDL Number: MFCD10687824
CAS Number: 1040684-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR